(R)-8-Fluoro-N-(1-oxo-1-(pyrrolidin-1-yl)-3-(3-(trifluoromethyl)phenyl)propan-2-yl)-1,2,3,4-tetrahydroisoquinoline-6-sulfonamide hydrochloride 98+%

Supplier: Ambeed
Warning

A382439-10MG A382439-1MG A382439-50MG A382439-5MG
77101-710EA 240.83 USD
77101-710 77101-712 77101-714 77101-716
(R)-8-Fluoro-N-(1-oxo-1-(pyrrolidin-1-yl)-3-(3-(trifluoromethyl)phenyl)propan-2-yl)-1,2,3,4-tetrahydroisoquinoline-6-sulfonamide hydrochloride 98+%
(R)-8-Fluoro-N-(1-oxo-1-(pyrrolidin-1-yl)-3-(3-(trifluoromethyl)phenyl)propan-2-yl)-1,2,3,4-tetrahydroisoquinoline-6-sulfonamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₆ClF₄N₃O₃S
MW: 535.98 g/mol
MDL Number: MFCD28133380
CAS Number: 1627607-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR