Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride 95+%

Supplier: Ambeed
Warning

Synonyms: trans-Methyl 6-methylpiperidine-3-carboxylate hydrochloride

A379963-100MG A379963-1G A379963-250MG A379963-5G
77101-676EA 63.66 USD
77101-676 77101-678 77101-680 77101-682
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride 95+%
Rel-Methyl (3R,6R)-6-methylpiperidine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂·ClH
MW: 193.671 g/mol
MDL Number: MFCD30181463
CAS Number: 1009376-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR