5-[4-(Trifluoromethoxy)phenyl]furan-2-carboxylic Acid 95%

Supplier: Ambeed
Warning

A379168-100MG A379168-1G A379168-250MG A379168-5G
77101-654EA 23 USD
77101-654 77101-656 77101-658 77101-660
5-[4-(Trifluoromethoxy)phenyl]furan-2-carboxylic Acid 95%
5-[4-(Trifluoromethoxy)phenyl]furan-2-carboxylic Acid

Precaución: for Research Use Only

Formula: C₁₂H₇F₃O₄
MDL Number: MFCD02215949
CAS Number: 638160-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR