tert-Butyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate 95%

Supplier: Ambeed
Warning

A378258-100MG A378258-1G A378258-250MG
77101-648EA 10 USD
77101-648 77101-650 77101-652
tert-Butyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate 95%
tert-Butyl 2-oxa-3-azabicyclo[2.2.1]hept-5-ene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₃
MDL Number: MFCD24465569
CAS Number: 99027-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR