N,N-Dipropargyl-p-toluenesulfonamide 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-N,N-di(prop-2-yn-1-yl)benzenesulphonamide

A377053-1G A377053-250MG A377053-5G
77101-638EA 127.31 USD
77101-638 77101-640 77101-642
N,N-Dipropargyl-p-toluenesulfonamide 98+%
N,N-Dipropargyl-p-toluenesulfonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂S
MW: 247.31 g/mol
MDL Number: MFCD22575206
CAS Number: 18773-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR