5-Fluoro-2-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Danger

A373072-25G A373072-5G
77101-610EA 69 USD
77101-610 77901-144
5-Fluoro-2-(trifluoromethyl)benzonitrile 98%
5-Fluoro-2-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₃F₄N
MDL Number: MFCD00236281
CAS Number: 240800-45-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR