5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamine dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Ceritinib dihydrochloride

A372261-10MG A372261-1MG A372261-50MG A372261-5MG A372261-100MG
77101-596EA 8.49 USD
77101-596 77101-598 77101-600 77101-602 77686-044
5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamine dihydrochloride 98%
5-Chloro-N2-(2-isopropoxy-5-methyl-4-(piperidin-4-yl)phenyl)-N4-(2-(isopropylsulfonyl)phenyl)pyrimidine-2,4-diamine dihydrochloride

Precaución: for Research Use Only

Formula: C₂₈H₃₈Cl₃N₅O₃S
MW: 631.06 g/mol
MDL Number: MFCD26142928
CAS Number: 1380575-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR