2-Bromo-5-iodoethylbenzene 95%

Supplier: Ambeed
Warning

A370919-100MG A370919-250MG
77101-578EA 10 USD
77101-578 77101-580
2-Bromo-5-iodoethylbenzene 95%
2-Bromo-5-iodoethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MDL Number: MFCD11846043
CAS Number: 1160573-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR