TAK-285 98+%

Supplier: Ambeed
Warning

Synonyms: N-(2-(4-((3-Chloro-4-(3-(trifluoromethyl)phenoxy)phenyl)amino)-5H-pyrrolo[3,2-d]pyrimidin-5-yl)ethyl)-3-hydroxy-3-methylbutanamide

A370749-10MG A370749-1MG A370749-50MG A370749-5MG
77101-570EA 55.17 USD
77101-570 77101-572 77101-574 77101-576
TAK-285 98+%
TAK-285

Precaución: for Research Use Only

Formula: C₂₆H₂₅ClF₃N₅O₃
MW: 548 g/mol
MDL Number: MFCD22124520
CAS Number: 871026-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR