4-Methoxyisobenzofuran-1,3-dione 97%

Supplier: Ambeed
Warning

A370481-1G A370481-5G A370481-100MG
77101-564EA 121 USD
77101-564 77101-566 77901-989
4-Methoxyisobenzofuran-1,3-dione 97%
4-Methoxyisobenzofuran-1,3-dione

Precaución: for Research Use Only

Formula: C₉H₆O₄
MDL Number: MFCD00034721
CAS Number: 14963-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR