2-Methylpyridine-3,4-diol 95%

Supplier: Ambeed
Warning

A369287-1G A369287-250MG
77101-548EA 10 USD
77101-548 77101-550
2-Methylpyridine-3,4-diol 95%
2-Methylpyridine-3,4-diol

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD01554236
CAS Number: 368838-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR