Pentafluorophenyl Diphenylphosphinate 95%

Supplier: Ambeed
Danger

Synonyms: Diphenylphosphinic acid Pentafluorophenyl ester, FDPP, Perfluorophenyl diphenylphosphinate

A368762-5G A368762-25G
77101-546EA 33.95 USD
77101-546 77594-094
Pentafluorophenyl Diphenylphosphinate 95%
Pentafluorophenyl Diphenylphosphinate

Precaución: for Research Use Only

Formula: C₁₈H₁₀F₅O₂P
MW: 384.24 g/mol
MDL Number: MFCD00192380
CAS Number: 138687-69-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR