(3-((Isopropoxycarbonyl)amino)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A368560-100MG A368560-1G A368560-250MG
77101-540EA 33 USD
77101-540 77101-542 77101-544
(3-((Isopropoxycarbonyl)amino)phenyl)boronic acid 98%
(3-((Isopropoxycarbonyl)amino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BNO₄
MDL Number: MFCD22375024
CAS Number: 1638329-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR