(2R,3S,5R)-5-Chloro-2-(((4-chlorobenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-chlorobenzoate 95%
Supplier: Ambeed
|
Warning
|
A368161-100G
A368161-10G
A368161-5G
77101-534EA
206.88
USD
77101-534
77101-536
77101-538
(2R,3S,5R)-5-Chloro-2-(((4-chlorobenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-chlorobenzoate 95%
(2R,3S,5R)-5-Chloro-2-(((4-chlorobenzoyl)oxy)methyl)tetrahydrofuran-3-yl 4-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₁₉H₁₅Cl₃O₅ MW: 429.67 g/mol |
MDL Number:
MFCD00672145 CAS Number: 21740-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...