4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine 97%

Supplier: Ambeed
Warning

A367645-25G A367645-5G
77101-530EA 74 USD
77101-530 77101-532
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine 97%
4-([1,1'-Biphenyl]-4-yl)-6-chloro-2-phenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅ClN₂
MDL Number: MFCD29477698
CAS Number: 1689538-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR