2,5-Dimethoxypyridine-4-boronic acid 95%
Supplier: Ambeed
|
Warning
|
A364938-250MG
A364938-100MG
A364938-5G
A364938-1G
77106-954EA
33
USD
77106-954
77686-032
77686-033
77901-571
2,5-Dimethoxypyridine-4-boronic acid 95%
2,5-Dimethoxypyridine-4-boronic acid
Precaución: for Research Use Only
Formula:
C₇H₁₀BNO₄ |
MDL Number:
MFCD18256800 CAS Number: 1630193-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...