N-(2,2-Dimethoxyethyl)-3-nitropyridin-2-amine 97%

Supplier: Ambeed
Warning

A364367-1G A364367-250MG A364367-5G
77106-948EA 55 USD
77106-948 77901-561 77900-630
N-(2,2-Dimethoxyethyl)-3-nitropyridin-2-amine 97%
N-(2,2-Dimethoxyethyl)-3-nitropyridin-2-amine

Precaución: for Research Use Only

Formula: C₉H₁₃N₃O₄
MDL Number: MFCD12068362
CAS Number: 1003947-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR