5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A360446-10G A360446-1G A360446-25G A360446-5G
77106-906EA 33.95 USD
77106-906 77106-908 77106-910 77106-912
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid 98%
5-Chloro-2-(methylthio)pyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂S
MW: 204.6300 g/mol
MDL Number: MFCD00173907
CAS Number: 61727-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR