(R)-4-Morpholino-1-(phenylthio)-2-butylamine 95%

Supplier: Ambeed
Warning

A360029-250MG A360029-50MG
77106-902EA 153 USD
77106-902 77106-904
(R)-4-Morpholino-1-(phenylthio)-2-butylamine 95%
(R)-4-Morpholino-1-(phenylthio)-2-butylamine

Precaución: for Research Use Only

Formula: C₁₄H₂₂N₂OS
MDL Number: MFCD17171370
CAS Number: 870812-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR