2-Methyl-3-morpholin-4-ylbenzoic acid 97%

Supplier: Ambeed
Warning

A359321-100MG A359321-1G A359321-250MG
77106-892EA 39 USD
77106-892 77106-894 77106-896
2-Methyl-3-morpholin-4-ylbenzoic acid 97%
2-Methyl-3-morpholin-4-ylbenzoic acid
Formula: C₁₂H₁₅NO₃
MDL Number: MFCD06740122
CAS Number: 886501-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR