2,4-Dichloronicotinc acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloropyridine-3-carboxylic acid, 2,4-Dichloro-3-pyridinecarboxylic acid, 2,4-Dichloronicotinic acid

A357901-100G A357901-10G A357901-1G A357901-25G A357901-5G
77106-870EA 345.86 USD
77106-870 77106-872 77106-874 77106-876 77106-878
2,4-Dichloronicotinc acid 98%
2,4-Dichloronicotinc acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD06796292
CAS Number: 262423-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR