2-Amino-2-(4-methylphenyl)ethan-1-ol 95%

Supplier: Ambeed
Warning

A357222-100MG A357222-1G
77106-858EA 91.24 USD
77106-858 77106-860
2-Amino-2-(4-methylphenyl)ethan-1-ol 95%
2-Amino-2-(4-methylphenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.20 g/mol
MDL Number: MFCD08277197
CAS Number: 157142-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR