(R)-2-Cyclopentylglycine 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Cyclopentyl-Gly-OH, (R)-2-Amino-2-cyclopentylacetic acid

A356166-100MG A356166-1G A356166-250MG A356166-5G A356166-25G
77106-848EA 10 USD
77106-848 77106-850 77106-852 77106-854 77900-497
(R)-2-Cyclopentylglycine 97%
(R)-2-Cyclopentylglycine

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MDL Number: MFCD03788075
CAS Number: 2521-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR