D-Leucine benzyl ester-p-toluenesulfonate 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Leu-OBzl.TosOH, H-D-Leu-OBzl·TosOH, H-D-Leu-OBzl.Tos-OH

A355378-10G A355378-5G
77106-844EA 29.71 USD
77106-844 77106-846
D-Leucine benzyl ester-p-toluenesulfonate 97%
D-Leucine benzyl ester-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₃H₁₉NO₂
MW: 393.50 g/mol
MDL Number: MFCD00066116
CAS Number: 17664-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR