2-Fluoro-4-iodo-5-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodo-5-picoline

A355116-1G A355116-250MG A355116-5G A355116-25G
77106-836EA 20.16 USD
77106-836 77106-838 77106-840 77686-010
2-Fluoro-4-iodo-5-methylpyridine 98%
2-Fluoro-4-iodo-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
MDL Number: MFCD03095297
CAS Number: 153034-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR