(3aR,5S,8aR,9aR)-5,8a-Dimethyl-3-methylene-3,3a,6,7,8,8a,9,9a-octahydronaphtho[2,3-b]furan-2(5H)-one 98%

Supplier: Ambeed
Warning

Synonyms: Alantolactone

A354359-10MG A354359-1MG A354359-25MG A354359-5MG A354359-100MG A354359-50MG
77106-826EA 22.28 USD
77106-826 77106-828 77106-830 77106-832 77686-008 77686-009
(3aR,5S,8aR,9aR)-5,8a-Dimethyl-3-methylene-3,3a,6,7,8,8a,9,9a-octahydronaphtho[2,3-b]furan-2(5H)-one 98%
(3aR,5S,8aR,9aR)-5,8a-Dimethyl-3-methylene-3,3a,6,7,8,8a,9,9a-octahydronaphtho[2,3-b]furan-2(5H)-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₂
MW: 232.31 g/mol
MDL Number: MFCD00046915
CAS Number: 546-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR