2-Mercapto-4,5-dimethylthiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dimethylthiazole-2(3H)-thione

A351059-100MG A351059-1G A351059-250MG
77106-790EA 22.28 USD
77106-790 77106-792 77106-794
2-Mercapto-4,5-dimethylthiazole 97%
2-Mercapto-4,5-dimethylthiazole

Precaución: for Research Use Only

Formula: C₅H₇NS₂
MW: 145.238 g/mol
MDL Number: MFCD00037895
CAS Number: 5351-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR