(R)-2-Amino-2-cyclopropylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Cyclopropyl-Gly-OH

A345319-100MG A345319-1G A345319-250MG A345319-5G
77106-728EA 19.1 USD
77106-728 77106-730 77106-732 77685-986
(R)-2-Amino-2-cyclopropylacetic acid 98%
(R)-2-Amino-2-cyclopropylacetic acid

Precaución: for Research Use Only

Formula: C₅H₉NO₂
MW: 115.13 g/mol
MDL Number: MFCD06659109
CAS Number: 49607-01-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR