3-Acetamidophenyl acetate 98%
Supplier: Ambeed
|
Warning
|
A345258-1G
A345258-250MG
77106-724EA
84.88
USD
77106-724
77106-726
3-Acetamidophenyl acetate 98%
3-Acetamidophenyl acetate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁NO₃ MW: 193.199 g/mol |
MDL Number:
MFCD00465580 CAS Number: 6317-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...