1-Bromo-3,5-dichlorobenzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dichlorobromobenzene
A344590-100G
A344590-10G
A344590-25G
A344590-500G
77106-716EA
36.08
USD
77106-716
77106-718
77106-720
77106-722
1-Bromo-3,5-dichlorobenzene 98%
1-Bromo-3,5-dichlorobenzene
Precaución: for Research Use Only
Formula:
C₆H₃BrCl₂ MW: 225.9 g/mol |
MDL Number:
MFCD00000584 CAS Number: 19752-55-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...