Methyl 2-Acetamido-2-phenylacetate 98%

Supplier: Ambeed
Warning

A344460-1G A344460-250MG A344460-5G
77106-710EA 106 USD
77106-710 77106-712 77106-714
Methyl 2-Acetamido-2-phenylacetate 98%
Methyl 2-Acetamido-2-phenylacetate

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MW: 207.226 g/mol
MDL Number: MFCD16691219
CAS Number: 36061-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR