2-Methyl-4-nitrophenylboronic acid 96%

Supplier: Ambeed
Warning

A344388-100MG A344388-1G A344388-250MG A344388-5G
77106-708EA 24 USD
77106-708 77594-018 77594-020 77612-968
2-Methyl-4-nitrophenylboronic acid 96%
2-Methyl-4-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BNO₄
MDL Number: MFCD18383626
CAS Number: 1228829-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR