Ethyl 3-cyanobenzoate 98%

Supplier: Ambeed
Warning

A344318-1G A344318-250MG A344318-5G
77106-702EA 43.5 USD
77106-702 77106-704 77106-706
Ethyl 3-cyanobenzoate 98%
Ethyl 3-cyanobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
MDL Number: MFCD03086010
CAS Number: 2463-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR