2-(4-(tert-Butoxycarbonyl)-2-oxopiperazin-1-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-N-Boc-2-oxo-piperazine-1-acetic acid

A343481-100MG A343481-1G A343481-5G
77106-688EA 28.65 USD
77106-688 77612-962 77685-980
2-(4-(tert-Butoxycarbonyl)-2-oxopiperazin-1-yl)acetic acid 98%
2-(4-(tert-Butoxycarbonyl)-2-oxopiperazin-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈N₂O₅
MW: 258.27 g/mol
MDL Number: MFCD04115297
CAS Number: 549506-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR