Boc-Dab(Z)-OH·DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Dicyclohexylamine (S)-4-(((benzyloxy)carbonyl)amino)-2-((tert-butoxycarbonyl)amino)butanoate

A341369-100G A341369-10G A341369-1G A341369-25G A341369-5G
77106-644EA 403 USD
77106-644 77106-646 77106-648 77106-650 77106-652
Boc-Dab(Z)-OH·DCHA 97%
Boc-Dab(Z)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₉H₄₇N₃O₆
MW: 533.71 g/mol
MDL Number: MFCD00236879
CAS Number: 16947-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR