6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine 95%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2,2-dimethyl-3,4-dihydro-2H-benzo[b][1,4]oxazine

A339836-100MG A339836-1G A339836-250MG
77106-616EA 13.8 USD
77106-616 77106-618 77106-620
6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine 95%
6-Chloro-2,2-dimethyl-3,4-dihydro-1,4-benzoxazine

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO
MW: 197.661 g/mol
MDL Number: MFCD14539604
CAS Number: 1216138-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR