4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-1H-1,2,3-triazole

A338425-100MG A338425-250MG A338425-50MG
77106-598EA 110.34 USD
77106-598 77106-600 77106-602
4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester 95%
4-(1H-1,2,3-Triazol-1-yl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BN₃O₂
MW: 271.123 g/mol
MDL Number: MFCD22570712
CAS Number: 1101174-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR