2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(3'-Chloro-[1,1'-biphenyl]-3-yl)-4,6-diphenyl-1,3,5-triazine

A338293-100MG A338293-250MG A338293-1G
77106-594EA 5.31 USD
77106-594 77106-596 77685-971
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine 95%
2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₇H₁₈ClN₃
MW: 419.91 g/mol
MDL Number: MFCD28975106
CAS Number: 1443049-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR