(2-Bromo-4-fluorophenyl)methanamine 95%

Supplier: Ambeed
Danger

A338185-25G A338185-1G
77106-592EA 229.16 USD
77106-592 77685-969
(2-Bromo-4-fluorophenyl)methanamine 95%
(2-Bromo-4-fluorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.03 g/mol
MDL Number: MFCD06213495
CAS Number: 739354-98-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR