Ethyl 3-Amino-4-hydroxybenzoate 95%

Supplier: Ambeed
Warning

A336014-100MG A336014-1G A336014-250MG A336014-5G
77106-582EA 11 USD
77106-582 77106-584 77106-586 77106-588
Ethyl 3-Amino-4-hydroxybenzoate 95%
Ethyl 3-Amino-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MDL Number: MFCD09961411
CAS Number: 13052-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR