4-(3-Chlorophenyl)-2,6-diphenylpyrimidine 98+%

Supplier: Ambeed
Warning

A334485-1G A334485-25G A334485-5G
77106-544EA 13.8 USD
77106-544 77106-546 77106-548
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine 98+%
4-(3-Chlorophenyl)-2,6-diphenylpyrimidine

Precaución: for Research Use Only

Formula: C₂₂H₁₅ClN₂
MW: 342.83 g/mol
MDL Number: MFCD29477687
CAS Number: 1536209-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR