(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene 95%

Supplier: Ambeed
Warning

A333173-250MG A333173-5G
77106-540EA 53.05 USD
77106-540 77106-542
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene 95%
(S)-6,6'-Dibromo-2,2'-dimethoxy-1,1'-binaphthalene

Precaución: for Research Use Only

Formula: C₂₂H₁₆Br₂O₂
MW: 472.169 g/mol
MDL Number: MFCD06654379
CAS Number: 117745-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR