4-Hydroxy-6-iodoquinoline 98%

Supplier: Ambeed
Warning

A332785-1G A332785-5G
77106-536EA 19 USD
77106-536 77106-538
4-Hydroxy-6-iodoquinoline 98%
4-Hydroxy-6-iodoquinoline

Precaución: for Research Use Only

Formula: C₉H₆INO
MW: 271.0545 g/mol
MDL Number: MFCD06808826
CAS Number: 342617-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR