Methyl-4-iodo-1H-pyrrole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-iodopyrrole-2-carboxylate

A331519-1G A331519-250MG A331519-25G A331519-5G
77099-984EA 46.68 USD
77099-984 77099-986 77099-988 77099-990
Methyl-4-iodo-1H-pyrrole-2-carboxylate 95%
Methyl-4-iodo-1H-pyrrole-2-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆INO₂
MW: 251.023 g/mol
MDL Number: MFCD07368824
CAS Number: 40740-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR