4-(Trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A329826-1G A329826-250MG A329826-25G A329826-5G
77099-974EA 14 USD
77099-974 77099-976 77099-978 77099-980
4-(Trifluoromethyl)pyridine 98%
4-(Trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄F₃N
MDL Number: MFCD00153289
CAS Number: 3796-24-5
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR