6-Bromo-3-chloro-2-methoxypyridine 96%

Supplier: Ambeed
Danger

A329810-100MG A329810-250MG
77099-970EA 46.68 USD
77099-970 77099-972
6-Bromo-3-chloro-2-methoxypyridine 96%
6-Bromo-3-chloro-2-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.46 g/mol
MDL Number: MFCD18257583
CAS Number: 1256810-58-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR