Methyl 3-Amino-2-hydroxypropanoate hydrochloride 98%

Supplier: Ambeed
Warning

A329278-1G A329278-5G
77099-964EA 14 USD
77099-964 77099-966
Methyl 3-Amino-2-hydroxypropanoate hydrochloride 98%
Methyl 3-Amino-2-hydroxypropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₄H₉NO₃·ClH
MDL Number: MFCD22741315
CAS Number: 186393-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR