2-(4-Chloro-2-methoxyphenyl)acetic acid 97%

Supplier: Ambeed
Warning

A328052-100MG A328052-250MG A328052-5G A328052-10G
77099-956EA 7 USD
77099-956 77099-958 77099-960 77612-888
2-(4-Chloro-2-methoxyphenyl)acetic acid 97%
2-(4-Chloro-2-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD09925126
CAS Number: 170737-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR