3-Iodo-DL-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(3-iodophenyl)propanoic acid, H-DL-Phe(3-I)-OH

A327367-1G A327367-250MG
77099-946EA 104 USD
77099-946 77099-948
3-Iodo-DL-phenylalanine 95%
3-Iodo-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀INO₂
MDL Number: MFCD05227974
CAS Number: 20846-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR