(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98%, ee 99%

Supplier: Ambeed
Warning

A326816-100MG A326816-250MG
77099-942EA 79.57 USD
77099-942 77099-944
(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine ≥98%, ee 99%
(11S,12S)-9,10-Dihydro-9,10-ethanoanthracene-11,12-diamine

Precaución: for Research Use Only

Formula: C₁₆H₁₆N₂
MW: 236.32 g/mol
MDL Number: MFCD06654768
CAS Number: 138517-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR